Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAMP2 Antibody (773816), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB376452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376452 Supplier Novus Biologicals Supplier No. FAB6500L
Add to Cart
Add to Cart

Rat Monoclonal Antibody

RAMP2 Monoclonal antibody specifically detects RAMP2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen RAMP2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 773816
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2
Host Species Rat
Immunogen E. coli-derived recombinant mouse RAMP2, Ser45-Val159, Accession # Q9WUP0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10266
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.