Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RanBP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1079814AC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1079814AC 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1079814AC Supplier Novus Biologicals Supplier No. NB10079814AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RanBP1 Polyclonal specifically detects RanBP1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen RanBP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HTF9A, MGC88701, RAN binding protein 1, Ran-binding protein 1, RanBP1, ran-specific GTPase-activating protein
Gene Symbols RANBP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human RAN Binding Protein 1 using the numbering given in entry NP_002873.1 (GeneID 5902).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5902
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.