Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RanBP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB436721 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436721 25 μL
NBP233881 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436721 Supplier Novus Biologicals Supplier No. NBP23388125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RanBP2 Polyclonal specifically detects RanBP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RanBP2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P49792
Gene Alias ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1
Gene Symbols RANBP2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SNSESLLGLLTSDKPLQGDGYSGAKPIPGGQTIGPRNTFNFGSKNVSGISFTENMGSSQQKNSGFRRSDDMFTFHGPGKSVFGTPTL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5903
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.