Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Reagent Additive 1

Catalog No. dy005
Click to view available options
Quantity:
21 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Heat-inactivated Normal Goat Serum, Diluted

1 vial (21mL) of diluted, heat-inactivated normal goat serum. Add 250μL of Reagent Additive 1 per 10mL of 1X detection antibody or reagent diluent.

Specifications

For Use With (Application) ELISA
Quantity 21 mL
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.