Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Reagent Diluent Concentrate 3 (5X)

Catalog No. dy004
Click to view available options
Quantity:
10 x 21 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

5X Concentrated 25% Tween 20 solution in PBS for DuoSets

10 vials (21mL/vial) of a 25% Tween 20 solution in PBS. The reagent diluent will require additional components for some DuoSets. Refer to the product-specific package. insert for more information.

Specifications

For Use With (Application) ELISA
Quantity 10 x 21 mL
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.