Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RanGAP1 Antibody (OTI1B4), PE, Novus Biologicals™
SDP

Catalog No. NBP273797PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273797PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273797PE Supplier Novus Biologicals Supplier No. NBP273797PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RanGAP1 Monoclonal specifically detects RanGAP1 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.

Specifications

Antigen RanGAP1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1B4
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias KIAA1835Fug1, MGC20266, Ran GTPase activating protein 1, ran GTPase-activating protein 1, RanGAP1, SD, segregation distorter homolog, segregation distorter homolog (Drosophila), segregation distortion
Gene Symbols RANGAP1
Host Species Mouse
Immunogen Full length recombinant protein of human RanGAP1 (NP_002874) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 5905.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.