Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANK/TNFRSF11A Antibody (044), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB079352 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB079352 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB079352 Supplier Novus Biologicals Supplier No. NBP290884FR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Rat samples. It is validated for ELISA.

Specifications

Antigen RANK/TNFRSF11A
Applications ELISA
Classification Monoclonal
Clone 44
Conjugate DyLight 680
Formulation 50 mM Sodium Borate
Gene Alias CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Rat RANK/TNFRSF11A (XP_573424.2; Met1-Pro213).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8792
Test Specificity No cross-reactivity in ELISA with: Rat EDAR
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.