Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANK/TNFRSF11A Antibody (107), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB333646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333646 Supplier Novus Biologicals Supplier No. NBP290324JF646
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human samples. It is validated for ELISA

Specifications

Antigen RANK/TNFRSF11A
Applications ELISA
Classification Monoclonal
Clone 107
Conjugate Janelia Fluor 646
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human RANK/TNFRSF11A (Accession#: NP_001257879.1, Met1-Pro212)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8792
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.