Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1A Antibody (RAP1 4C8/1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250269AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250269AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250269AI Supplier Novus Biologicals Supplier No. NBP250269AF700

Mouse Monoclonal Antibody

RAP1A Monoclonal specifically detects RAP1A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen RAP1A
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone RAP1 4C8/1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
Gene Symbols RAP1A
Host Species Mouse
Immunogen Human Rap1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Endosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 5906
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.