Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1B Antibody, Novus Biologicals™
SDP

Catalog No. NBP220072 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220072 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220072 Supplier Novus Biologicals Supplier No. NBP220072
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAP1B Polyclonal specifically detects RAP1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RAP1B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P61224
Gene Alias DKFZp586H0723, GTP-binding protein smg p21B, K-REV, RAL1B, RAP1B, member of RAS oncogene family, Ras family small GTP binding protein RAP1B, ras-related protein Rap-1b, RAS-related protein RAP1B, small GTP binding protein
Gene Symbols RAP1B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAP1B. The exact sequence is proprietary.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endosome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5908
Test Specificity Human. Species reactivity based on sequence identity: Rhesus Monkey(100%), Chicken (100%), Chimpanzee (100%), Bovine (100%), Mouse (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.