Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1GAP Antibody, Novus Biologicals™
SDP

Catalog No. NBP15307220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15307220 20 μL
NBP153072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15307220 Supplier Novus Biologicals Supplier No. NBP15307220UL

Rabbit Polyclonal Antibody

RAP1GAP Polyclonal specifically detects RAP1GAP in Human samples. It is validated for Western Blot, SDS-Page.

Specifications

Antigen RAP1GAP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P47736
Gene Alias GTPase activating protein 1, RAP1 GTPase activating protein
Gene Symbols RAP1GAP
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RAP1GAP(RAP1 GTPase activating protein) The peptide sequence was selected from the middle region of RAP1GAP. Peptide sequence IENIQEVQEKRESPPAGQKTPDSGHVSQEPKSENSSTQSSPEMPTTKNRA.
Molecular Weight of Antigen 73 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Lysosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 5909
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.