Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP2A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7205499 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9007 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9007 Supplier Proteintech Group Inc Supplier No. 137891AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAP2A is a signaling protein involved in small GTPase mediated signal transduction activated by guanine nucleotide-exchange factors (GEF) EPAC and EPAC2, in a cAMP-dependent manner, and GFR. This membrane protein belongs to the small GTPase superfamily and Ras family and contains domains required for GTP binding and a Ras-like C-terminal motif for post-translational lipid binding and subsequent anchoring to the plasma membrane. Apart from having a possible role in membrane shedding, RAP2A also functions as a slowly responding molecular switch in the RAP1A signaling cascade and also interacts with ARHGAP29, the putative specific effector of RAP2A.

Specifications

Antigen RAP2A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAP2A
Gene Accession No. P10114, Q80ZJ1
Gene Alias K REV, KREV, RAP2, RAP2A, Ras related protein Rap 2a, RbBP 30
Gene Symbols RAP2A
Host Species Rabbit
Immunogen RAP2A Fusion Protein Ag4808
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5911, 76108
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.