Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP80 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292388 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292388 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292388 Supplier Novus Biologicals Supplier No. NB1002346PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAP80 Polyclonal specifically detects RAP80 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen RAP80
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1
Host Species Rabbit
Immunogen Maps to a region between residue 600 and 650 of human Receptor Associated Protein 80 using the numbering given in entry NP_057374.3 (GeneID 51720).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51720
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.