Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rapamycin
GREENER_CHOICESDP

Catalog No. 12921 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-921 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-921 Supplier Tocris Bioscience™ Supplier No. 1292/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

mTOR inhibitor; immunosuppressant

Rapamycin is a antifungal and immunosuppressant. Specific inhibitor of mTOR (mammalian target of Rapamycin). Complexes with FKBP-12 and binds mTOR inhibiting its activity. Inhibits interleukin-2-induced phosphorylation and activation of p70 S6 kinase. Induces autophagy in yeast and mammalian cell lines. Drives hPSC differentiation to mesendoderm and blood progenitor cells. Also used as a chemical dimerizer; rapamycin and GA3-AM (Cat. No. 5407) chemically inducible dimerization systems are orthogonal. Inhibits MERS-CoV infection of Huh7 cells in a plaque reduction assay. Enhances lentiviral transduction of hematopoietic stem cells.

Chemical Identifiers

CAS 53123-88-9
Molecular Formula C51H79NO13

Specifications

Quantity 1 mg
Formula Weight 914.18
Product Type Rapamycin
Purity 98%
Molecular Weight (g/mol) 914.187
Chemical Name or Material Rapamycin
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.