Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP106549G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106549G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106549G Supplier Novus Biologicals Supplier No. NBP106549G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAPGEF2 Polyclonal specifically detects RAPGEF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RAPGEF2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CNrasGEF, KIAA0313, Neural RAP guanine nucleotide exchange protein, nRap GEP, NRAPGEP, PDZ domain containing guanine nucleotide exchange factor (GEF) 1, PDZGEF1, PDZ-GEF1PDZ domain-containing guanine nucleotide exchange factor 1, RA(Ras/Rap1A-associating)-GEF, RA-GEFDKFZP586O1422, Rap guanine nucleotide exchange factor (GEF) 2, rap guanine nucleotide exchange factor 2, Rap-GEP
Gene Symbols RAPGEF2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 975 and 1025 of human PDZ domain containing guanine nucleotide exchange factor (GEF) 1 (NP_055062.1).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9693
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.