Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF3 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB165005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165005 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB165005 Supplier Novus Biologicals Supplier No. NBP281767FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAPGEF3 Polyclonal antibody specifically detects RAPGEF3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RAPGEF3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias 9330170P05Rik, bcm910, cAMP-GEFI, cAMP-regulated guanine nucleotide exchange factor I, CGEF1, EPAC 1, EPAC1, EPACRAP guanine-nucleotide-exchange factor (GEF) 3, Exchange factor directly activated by cAMP 1, Exchange protein directly activated by cAMP 1, HSU79275, MGC21410, Rap guanine nucleotide exchange factor (GEF) 3, rap guanine nucleotide exchange factor 3, Rap1 guanine-nucleotide-exchange factor directly activated by cAMP
Host Species Rabbit
Immunogen RAPGEF3 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human RAPGEF3. The immunogen is located within amino acids 60 - 110 of RAPGEF3. Amino Acid Squence: WTPLTNSEESLDFSESLE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10411
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.