Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPH1 Antibody, Novus Biologicals™
SDP

Catalog No. NB173793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173793 Supplier Novus Biologicals Supplier No. NBP318515
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAPH1 Polyclonal antibody specifically detects RAPH1 in Human samples. It is validated for Immunoprecipitation

Specifications

Antigen RAPH1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Immunoprecipitation 8 - 15 ug/mg lysate
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias candidate 18, candidate 9, lamellipodin, PREL2, PREL-2, proline rich EVH1 ligand 2, Protein RMO1, Ras association (RalGDS/AF-6) and pleckstrin homology domains 1, ras-associated and pleckstrin homology domains-containing protein 1, RMO1
Host Species Rabbit
Immunogen Recognizes region between residues Between Between 1200 and 1250..
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 65059
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.