Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Raptor™ HILIC-Si LC Columns, Particle Size: 2.7 μm
SDP

Catalog No. 06611456 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
06-611-456 30 mm 2.1 mm
06-611-457 50 mm 2.1 mm
06-611-459 50 mm 3 mm
06-611-458 50 mm 4.6 mm
06-611-453 100 mm 2.1 mm
06-611-455 100 mm 3 mm
06-611-454 100 mm 4.6 mm
06-611-460 150 mm 2.1 mm
06-611-462 150 mm 3 mm
06-611-461 150 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 06-611-456 Supplier Restek™ Supplier No. 9310A32
Add to Cart
Add to Cart

Designed for increased retention of polar compounds without ion-pairing reagents

  • 2.7 μm Raptor core-shell particles provide the speed of SPP, with greater efficiency and capacity than 5 μm particles
  • Fully compatible with both HPLC and UHPLC when using low back-pressure HILIC mobile phases
  • Switch to a Raptor HILIC-Si LC column when more retention and resolution of hydrophilic, polar analytes is needed
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 30 mm
Max. Pressure 600 bar (8,700 psi)
Particle Size 2.7 μm
pH 2 to 8
Pore Size 90 Å
Surface Area 130 m2/g
Temperature Max: 80°C
USP Type L13
Stationary Phase Bare Silica
Packing Material Superficially Porous Silica (SPP or Core-Shell)
Product Type Column
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.