Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAR alpha/NR1B1 Antibody (OTI2C3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273802IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273802IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273802IR Supplier Novus Biologicals Supplier No. NBP273802IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAR alpha/NR1B1 Monoclonal specifically detects RAR alpha/NR1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RAR alpha/NR1B1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C3
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha
Gene Symbols RARA
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 52-299 of human RAR alpha/NR1B1 (NP_000955) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Phospho Specific, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5914.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.