Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Ras Activation ELISA ASSAY Kit
Click to view available options
Quantity:
96 Assays
Description
Includes
- Glutathione-Coated Plate, 96-well strip format
- Anti-Ras Detection Antibody
- Mg2+ Lysis/Wash Buffer, 5X
- 20% Tween™ 20
- 10% BSA in TBS (Blocking Buffer)
- 20X TBS Wash
- Gt x Ms, HRP
- Chemiluminescent Reaction Buffer
- Chemiluminescent Detection Reagent
- Store at -80º C
- Raf-1-RBD, GST
- EGF stimulated HeLa Cell Lysate
Requires
- Multi-channel or repeating pipettes
- Plate shaker at room temperature and at 4oC.
- Pipettors and tips capable of accurately measuring 1-1000μL
- Graphing software for plotting data or graph paper for manual plotting of data
- Multi-channel pipettor reservoirs
- Protease inhibitors
- Luminometer or CCD camera coupled imaging system
Applications: ELISA, Cell Function Assay

Specifications
Specifications
For Use With (Application) | ELISA, Cell Function Assay |
Quantity | 96 Assays |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction