Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ras Rabbit anti-Human, Clone: JF10-11, Novus Biologicals™
SDP

Catalog No. NBP267097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267097 Supplier Novus Biologicals Supplier No. NBP267097

Rabbit Monoclonal Antibody

Ras Monoclonal specifically detects Ras in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Ras
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone JF10-11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1:10 - 1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Symbols KRAS
Host Species Rabbit
Immunogen Synthetic peptide within Human Ras aa 21-64/ 189. (SwissProt: P01111 Human; SwissProt: P01112 Human; SwissProt: P01116 Human; SwissProt: P08556 Mouse; SwissProt: P32883 Mouse; SwissProt: Q61411 Mouse; SwissProt: P08644 Rat; SwissProt: P20171 Rat; SwissProt: Q04970 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.