Learn More
Invitrogen™ RASA1 Recombinant Rabbit Monoclonal Antibody (JE60-89)

Description
RASA1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RASA1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | JE60-89 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | RASA1 |
| Gene Accession No. | P20936 |
| Gene Alias | CMAVM; CM-AVM; Gap; GAPX; GTPase-activating protein; p120; p120 RAS GTPase activating protein; p120GAP; p120RASGAP; p120-rasGAP; PKWS; Ras GTPase-activating protein 1; ras GTPase-activating protein 1-like protein; Ras p21 protein activator; RAS p21 protein activator (GTPase activating protein RAS p21); RAS p21 protein activator (GTPase activating protein) 1; RAS p21 protein activator 1; Rasa; RASA1; RASGAP; triphosphatase-activating protein; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.