Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Rasagiline mesylate
GREENER_CHOICESDP

Catalog No. 430850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-085-0 Supplier Tocris Bioscience™ Supplier No. 4308/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, irreversible MAO-B inhibitor

Selective irreversible MAO-B inhibitor. Displays antiParkinsonian activity; exhibits neuroprotective and antiapoptotic activity against several neurotoxins in cell culture.

Chemical Identifiers

CAS 161735-79-1
Molecular Formula C13H17NO3S

Specifications

Quantity 50 mg
Formula Weight 267.34
Purity >99%
Molecular Weight (g/mol) 267.34
Chemical Name or Material Rasagiline mesylate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.