Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGEF1C Antibody, Novus Biologicals™
SDP

Catalog No. NBP158874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP158874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP158874 Supplier Novus Biologicals Supplier No. NBP158874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RASGEF1C Polyclonal specifically detects RASGEF1C in Human samples. It is validated for Western Blot.

Specifications

Antigen RASGEF1C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N431
Gene Alias FLJ35841, RasGEF domain family, member 1C, ras-GEF domain-containing family member 1C
Gene Symbols RASGEF1C
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RASGEF1C(RasGEF domain family, member 1C) The peptide sequence was selected from the middle region of RASGEF1C. Peptide sequence FLELAKQVGEFITWKQVECPFEQDASITHYLYTAPIFSEDGLYLASYESE.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 255426
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Equine: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Chicken: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.