Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGRF2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220087 Supplier Novus Biologicals Supplier No. NBP220087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RASGRF2 Polyclonal specifically detects RASGRF2 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RASGRF2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. O14827
Gene Alias DKFZp781H1715, GRF2RAS-GRF2, guanine nucleotide-releasing factor 2, Ras guanine nucleotide exchange factor 2, Ras protein-specific guanine nucleotide-releasing factor 2, Ras-GRF2, ras-specific guanine nucleotide-releasing factor 2
Gene Symbols RASGRF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RASGRF2. The exact sequence is proprietary.
Molecular Weight of Antigen 141 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5924
Test Specificity Mouse. Species reactivity based on sequence identity: Human (100%).
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.