Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASL10A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158920
Description
RASL10A Polyclonal specifically detects RASL10A in Human samples. It is validated for Western Blot.Specifications
RASL10A | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
ras-like protein family member 10A, RAS-like, family 10, member A, RAS-related on chromosome 22, Ras-related protein on chromosome 22, RRP22Ras-like protein RRP22 | |
Rabbit | |
23 kDa | |
100 μL | |
Signal Transduction | |
10633 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q92737 | |
RASL10A | |
Synthetic peptides corresponding to RASL10A(RAS-like, family 10, member A) The peptide sequence was selected from the N terminal of RASL10A (NP_006468). Peptide sequence PTDGPRLYRPAVLLDGAVYDLSIRDGDVAGPGSSPGGPEEWPDAKDWSLQ. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Equine: 85%. | |
Human, Equine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction