Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF1 Antibody (3F3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP104342R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104342R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104342R Supplier Novus Biologicals Supplier No. NBP104342R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RASSF1 Monoclonal specifically detects RASSF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RASSF1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3F3
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5
Gene Symbols RASSF1
Host Species Mouse
Immunogen Recombinant human RASSF1A (1-340 aa), purified from E. coli (NP_009113).
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 11186
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.