Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF1 Antibody (3F3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310185 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310185 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310185 Supplier Novus Biologicals Supplier No. NBP104342MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RASSF1 Monoclonal antibody specifically detects RASSF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen RASSF1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3F3
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5
Host Species Mouse
Immunogen Recombinant human RASSF1 (1-340 aa), purified from E. coli (NP_009113).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 11186
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.