Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF2 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB045696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045696 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045696 Supplier Novus Biologicals Supplier No. NBP297615AF594

Rabbit Polyclonal Antibody

RASSF2 Polyclonal specifically detects RASSF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen RASSF2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0168DKFZp781O1747, Ras association (RalGDS/AF-6) domain family 2, Ras association (RalGDS/AF-6) domain family member 2, ras association domain-containing protein 2, RASFADIN
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RASSF2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9770
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.