Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP120967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120967 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120967 Supplier Novus Biologicals Supplier No. NBP120967
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RASSF3 Polyclonal specifically detects RASSF3 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen RASSF3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_835463.1
Gene Alias MGC119194, MGC119195, MGC119197, Ras association (RalGDS/AF-6) domain family 3, Ras association (RalGDS/AF-6) domain family member 3, ras association domain-containing protein 3, RASSF5
Gene Symbols RASSF3
Host Species Goat
Immunogen Peptide with sequence CKPPQTSPNSGK corresponding to C-Terminus according to NP_835463.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 283349
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.