Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF8 Antibody (OTI1F5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP273820G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273820G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273820G Supplier Novus Biologicals Supplier No. NBP273820G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RASSF8 Monoclonal specifically detects RASSF8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RASSF8
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F5
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C12orf2, carcinoma associated HOJ-1, Carcinoma-associated protein HOJ-1, chromosome 12 open reading frame 2, DKFZp434O0227, FLJ11542, HoJ-1, HOJ1, Ras association (RalGDS/AF-6) domain family (N-terminal) member 8, ras association domain-containing protein 8
Gene Symbols RASSF8
Host Species Mouse
Immunogen Full length recombinant protein of human RASSF8 (NP_009142) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 11228.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.