Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat alpha 2u-Globulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB586SP
This item is not returnable.
View return policy
Description
alpha 2u-Globulin Monoclonal specifically detects alpha 2u-Globulin in Rat samples. It is validated for Immunohistochemistry.Specifications
alpha 2u-Globulin | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
259246 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunohistochemistry | |
129736 | |
Immunohistochemistry 8-25 ug/mL | |
AUG, Mup, PGCL1 | |
Rat urine-derived alpha 2u -Globulin | |
25 μg | |
Primary | |
Detects rat alpha 2μ -Globulin in direct ELISAs. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction