Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat alpha 2u-Globulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | alpha 2u-Globulin |
---|---|
Clone | 129736 |
Dilution | Immunohistochemistry 8-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Description
alpha 2u-Globulin Monoclonal specifically detects alpha 2u-Globulin in Rat samples. It is validated for Immunohistochemistry.Specifications
alpha 2u-Globulin | |
Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Rat | |
AUG, Mup, PGCL1 | |
Rat urine-derived alpha 2u -Globulin | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat alpha 2μ -Globulin in direct ELISAs. |
129736 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
259246 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title