Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-CD43/Sialophorin, Clone: R2/60, FITC, Novus Biologicals™
SDP

Catalog No. NBP1440115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1440115 0.5 mg
NBP14401105 0.05 mg
NBP14401101 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP1440115 Supplier Novus Biologicals Supplier No. NBP1440110.5MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

CD43/Sialophorin Monoclonal specifically detects CD43/Sialophorin in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD43/Sialophorin
Applications Flow Cytometry
Classification Monoclonal
Clone R2/60
Conjugate FITC
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Alias CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
Gene Symbols SPN
Host Species Rat
Immunogen The immunogen for this antibody was CD43.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6693
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.