Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Ly-6G6C, Clone: NIMP-R14, FITC, Novus Biologicals™
SDP

Catalog No. NBP16013905 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16013905 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP16013905 Supplier Novus Biologicals Supplier No. NBP1601390.05MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 8 publications

Ly-6G6C Monoclonal specifically detects Ly-6G6C in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoassay.

Specifications

Antigen Ly-6G6C
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone NIMP-R14
Concentration 0.1 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay
Formulation 1 ml (100 ug/ml) 0.2 um filtered FITC conjugated antibody solution in PBS and 1.0% BSA with 0.05% Sodium Azide
Gene Alias C6orf24, G6c, lymphocyte antigen 6 complex locus protein G6c, lymphocyte antigen 6 complex, locus G6C, NG24, UNQ1947/PRO4430
Gene Symbols LY6G6C
Host Species Rat
Immunogen Purified BALB/c mouse neutrophils
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80740
Test Specificity mouse Ly-6G6C
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.