Learn More
Description
Specifications
Specifications
| Antigen | MHC Class II |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M5/114.15.2 |
| Concentration | 0.2 mg/mL |
| Conjugate | PE |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Formulation | Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide |
| Gene Alias | class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM |
| Gene Symbols | HLA-DMA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
