Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | CX3CL1/Fractalkine |
---|---|
Clone | 96834 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Rat samples. It is validated for Western Blot.Specifications
CX3CL1/Fractalkine | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
CX3CL1 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
96834 | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
O55145 | |
6376 | |
E. coli-derived recombinant rat CX3CL1/Fractalkine Chemokine Domain Gln25-Gly100 Accession # O55145 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat CX3CL1/Fractalkine Chemokine in direct ELISAs and Western blots. In ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) CX3CL1 is observed, 10% cross-reactivity with rmCCL11 is observed, and no cross-reactivity with recombinant human (rh) CX3CL1 or rhCCL11 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title