Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | CXCL1/GRO alpha/KC/CINC-1 |
---|---|
Clone | 251021 |
Dilution | Western Blot 1 ug/mL, Neutralization 3-15 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
CXCL1/GRO alpha/KC/CINC-1 Monoclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Rat samples. It is validated for Western Blot, Neutralization.Specifications
CXCL1/GRO alpha/KC/CINC-1 | |
Western Blot 1 ug/mL, Neutralization 3-15 ug/mL | |
Monoclonal | |
Mouse | |
Rat | |
P14095 | |
2919 | |
E. coli-derived recombinant rat CXCL1/GRO alpha/KC/CINC-1 Ala25-Lys96 Accession # P14095 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat CXCL1/GRO alpha/KC/CINC-1 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) GRO alpha , rhGRO beta , rhGRO gamma , and recombinant mouse KC is observed. No cross-reactivity with recombinant rat (rr) CINC2a, rrCXCL3, rhCXCL5, 6, 7, 8, 9, 10, 11, 12 or 13, rmCXCL2, 6, 9, 10, 12, 13, or recombinant porcine CXCL8 is observed. |
251021 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
Cxcl1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title