Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Dkk-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Dkk-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Dkk-1 Polyclonal specifically detects Dkk-1 in Rat samples. It is validated for Western Blot.Specifications
Dkk-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Rat | |
dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
DKK1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_001099820 | |
22943 | |
Mouse myeloma cell line NS0-derived recombinant rat Dkk-1 (R&D Systems, Catalog # 4010-DK) Ser23-His270 Accession # NP_001099820 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat Dkk-1 in direct ELISAs and Western blots. In these formats, approximately 40% cross-reactivity with recombinant mouse Dkk-1 is observed and approximately 20% cross-reactivity with recombinant human Dkk-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title