Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat HMGB1/HMG-1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB183280 Shop All Novus Biologicals Products
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 31.25 to 2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA
Gene ID (Entrez) 3146
Gene Symbol HMGB1
Quantity 1 Kit
Research Discipline Cellular Markers, Chromatin Research, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Target Rat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.