Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat ICAM-1/CD54 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5832100
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
ICAM-1/CD54 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3383 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
ELISA | |
141017 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q00238 | |
ICAM1 | |
Mouse myeloma cell line NS0-derived recombinant rat ICAM-1 Met1-Thr493 Accession # Q00238 | |
100 μg | |
Primary | |
Detects rat ICAM-1 in ELISAs and Western blots. Does not cross-react with recombinant human (rh) CD31, recombinant mouse (rm) DCC, rhICAM-1, rmICAM-1, rmICAM-2, rhICAM-3, rhMAdCAM-1, or rmVCAM-1. | |
Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction