Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat ICAM-1/CD54 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $1798.00

Specifications

Antigen ICAM-1/CD54
Clone 141017
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5832SP
GREENER_CHOICE
View Documents
R&D Systems
MAB5832SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB5832100
GREENER_CHOICE
View Documents
R&D Systems
MAB5832100
100 μg
Each for $491.00
Only null left
Add to Cart
 
MAB5832500
GREENER_CHOICE
View Documents
R&D Systems
MAB5832500
500 μg
Each for $1,798.00
Only null left
Add to Cart
 
Description

Description

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

ICAM-1/CD54
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Rat
Q00238
3383
Mouse myeloma cell line NS0-derived recombinant rat ICAM-1 Met1-Thr493 Accession # Q00238
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat ICAM-1 in ELISAs and Western blots. Does not cross-react with recombinant human (rh) CD31, recombinant mouse (rm) DCC, rhICAM-1, rmICAM-1, rmICAM-2, rhICAM-3, rhMAdCAM-1, or rmVCAM-1.
141017
ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
ICAM1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.