Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IFN-gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB585100
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Rat samples. It is validated for Western Blot, Neutralization.Specifications
| IFN-gamma | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3458 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization | |
| 88915 | |
| Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL | |
| P01581 | |
| IFNG | |
| E. coli-derived recombinant rat IFN-gamma Gln23-Cys156 Accession # P01581 | |
| 100 μg | |
| Primary | |
| Detects rat IFN-gamma in Western blots. In Western blots, this antibody shows less than 1% cross-reactivity with recombinant mouse (rm) IFN-gamma and no cross-reactivity with rhIFN-gamma . | |
| Rat | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction