Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IFN-gamma Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $1929.00

Specifications

Antigen IFN-gamma
Clone 88915
Dilution Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL
Applications Western Blot, Neutralization
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB585SP
GREENER_CHOICE
View Documents
R&D Systems
MAB585SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB585100
GREENER_CHOICE
View Documents
R&D Systems
MAB585100
100 μg
Each for $526.00
Only null left
Add to Cart
 
MAB585500
GREENER_CHOICE
View Documents
R&D Systems
MAB585500
500 μg
Each for $1,929.00
Only null left
Add to Cart
 
Description

Description

IFN-gamma Monoclonal specifically detects IFN-gamma in Rat samples. It is validated for Western Blot, Neutralization.
Specifications

Specifications

IFN-gamma
Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL
Monoclonal
Mouse
Rat
P01581
3458
E. coli-derived recombinant rat IFN-gamma Gln23-Cys156 Accession # P01581
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rat IFN-gamma in Western blots. In Western blots, this antibody shows less than 1% cross-reactivity with recombinant mouse (rm) IFN-gamma and no cross-reactivity with rhIFN-gamma .
88915
Western Blot, Neutralization
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
IFNG
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.