Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b Isotype Control (LF2), Novus Biologicals™
SDP

Catalog No. NB004941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004941 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004941 Supplier Novus Biologicals Supplier No. NBP221941250UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Rat samples. It is validated for .

Specifications

Antigen IgG2b
Classification Monoclonal
Clone LF2
Concentration LYOPH
Conjugate Unconjugated
Formulation 0.2 um filtered solution in PBS.
Host Species Rat
Purification Method Protein A or G purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Reconstitution Reconstitute with 1.0 ml PBS.
Target Species Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.