Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat IL-6 ELISA Kit (Colorimetric)
SDP

Catalog No. nbp192697 Shop All R&D Systems Products
Click to view available options
Quantity:
96 Tests

Catalog No. NBP192697

Supplier: Novus Biologicals™ NBP192697

Only null left
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Rat IL-6 ELISA Kit (Colorimetric) measures rat IL-6 in cell culture supernatant, serum.

Specifications

Assay Range 31.0 to 2,000 pg/mL
Assay Sensitivity 12.0 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Serum, Serum
Specificity The interference of circulating factors of the immune system was evaluated by spiking these proteins at physiologically relevant concentrations into a rat IL-6 positive serum. There was no crossreactivity detected.
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 3569
Gene Symbol IL6
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-6, Biotin-Conjugate anti-rat IL-6 monoclonal antibody, Streptavidin-HRP, Rat IL-6 Standard lyophilized, 4ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Research Discipline Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6
Target IL-6
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.