Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat LILRC1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4074SP
This item is not returnable.
View return policy
Description
LILRC1 Polyclonal specifically detects LILRC1 in Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
LILRC1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Lilra5 | |
Mouse myeloma cell line NS0-derived recombinant rat LILRC1 Gln17-Asn248 Accession # NP_001070261 | |
25 μg | |
Primary | |
Detects rat LILRC1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse LILRCI is observed and less than 1% cross-reactivity with recombinant rat LILRC2 is observed. | |
Rat | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
NP_001070261 | |
Goat | |
Affinity Purified | |
RUO | |
232801 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction