Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat LILRC1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | LILRC1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Description
LILRC1 Polyclonal specifically detects LILRC1 in Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
LILRC1 | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
232801 | |
Mouse myeloma cell line NS0-derived recombinant rat LILRC1 Gln17-Asn248 Accession # NP_001070261 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat LILRC1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse LILRCI is observed and less than 1% cross-reactivity with recombinant rat LILRC2 is observed. |
LYOPH | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
NP_001070261 | |
Lilra5 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title