Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Pentraxin 2/SAP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $643.50
Specifications
Antigen | Pentraxin 2/SAP |
---|---|
Clone | 273902 |
Concentration | LYOPH |
Dilution | Blockade of Receptor-ligand Interaction |
Applications | Blockade of Receptor-ligand Interaction |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB18951SP
![]() |
R&D Systems
MAB18951SP |
25 μg |
Each for $200.50
|
|
|||||
MAB18951
![]() |
R&D Systems
MAB18951 |
500 μg |
Each for $643.50
|
|
|||||
Description
Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
Pentraxin 2/SAP | |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Mouse | |
Rat | |
P23680 | |
325 | |
Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 (predicted) Accession # P23680 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat Pentraxin 2/SAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Pentraxin 2 or recombinant mouse Pentraxin 2 is observed. |
273902 | |
Blockade of Receptor-ligand Interaction | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component | |
APCS | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title