Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat RAGE Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF1616SP
This item is not returnable.
View return policy
Description
AGER Polyclonal specifically detects AGER in Rat samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Specifications
RAGE | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
Goat | |
Affinity Purified | |
RUO | |
5891 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q63495 | |
Ager | |
Mouse myeloma cell line NS0-derived recombinant rat RAGE Gln24-Ala342 Accession # Q63495 | |
25 μg | |
Primary | |
Detects rat RAGE in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human RAGE and recombinant canine RAGE is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction